Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
41807-FF
41807-FI
CAAA41807-FFEA
1106
CAD
CAAA41807-FF
CAAA41807-FI
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Platinum Iridium alloy (80:20 w%)
MDL: MFCD02091683
Purity: 99.9%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈1.36g/25x25mm
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...