Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
41802-FF
41802-FI
41802-GH
CAAA41802-FFEA
521.4
CAD
CAAA41802-FF
CAAA41802-FI
CAAA41802-GH
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Platinum Iridium alloy (90:10 w%)
MDL: MFCD02091683
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.34g/25x25mm
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...