Aluminium ≥99.0% (metals basis), thin foil, Thickness: 10.0 micron (0.0004 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

41798-FI 41798-GH
CAAA41798-FIEA 79.95 CAD
CAAA41798-FI CAAA41798-GH
Aluminium ≥99.0% (metals basis), thin foil, Thickness: 10.0 micron (0.0004 in)
Aluminium

MDL: MFCD00134029


Purity: 99.0% CAS No.: 7429-90-5 Physical Form: ≈0.067g/50x50mm, not certified pinhole free Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR