3,4-Difluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

41789-03 41789-06
CAAA41789-03EA 52.46 CAD
CAAA41789-03 CAAA41789-06 CAAA41789-14
3,4-Difluorotoluene 98%
3,4-Difluorotoluene

MDL: MFCD00075087


Purity: 98% CAS No.: 2927-34-6 Molecular Formula: C7H6F2 Formula Weight: 128.12 Flash Point: 25°(77°F) Physical Form: Liquid Density: 1.120 Refractive Index: 1.4500 Odor: Aromatic Boiling Point: 110-113° UN No.: 1993 MDL No.: MFCD00075087

Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 110…113 °C
Density: 1.120
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075087
CAS Number: 2927-34-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR