Sodium tellurite(IV) ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
41778-09
41778-18
CAAA41778-09EA
94.17
CAD
CAAA41778-09
CAAA41778-18
Sodium tellurite(IV) ≥99.5% (metals basis)
Sodium tellurite(IV)
MDL: MFCD00036141
Soluble in water
Purity: 99.5%
CAS No.: 10102-20-2
Molecular Formula: Na2O3Te
Formula Weight: 221.58
Physical Form: -100 Mesh Powder
Appearance: White
Solubility: Soluble in water
Merck Reference: 14,8685
UN No.: 3284
MDL No.: MFCD00036141
Formula:
Na₂O₃Te Melting Pt: 710 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036141 CAS Number: 10102-20-2 EINECS: 233-268-4 UN: 3284 ADR: 6.1,II Merck Index: 14,8685 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...