Phthalic anhydride, anhydrous 99.0-100.2% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
41771-36
41771-A4
CAAA41771-36EA
108.17
CAD
CAAA41771-36
CAAA41771-A4
Phthalic anhydride, anhydrous 99.0-100.2% ACS
Phthalic anhydride
MDL: MFCD00005918
Beilstein Registry No.: 118515
Fieser: 1,882 21,351
Purity: 99.0-100.2%
CAS No.: 85-44-9
Molecular Formula: C6H4(CO)2O
Formula Weight: 148.12
Flash Point: 152°(305°F)
Physical Form: Flaky Crystals
Density: 1.53
Sensitivity: Moisture Sensitive
Odor: Weak, characteristic
Melting Point: 131-133°
Boiling Point: 295° subl.
Beilstein Reference: 118515
Merck Reference: 14,7372
UN No.: 2214
MDL No.: MFCD00005918
Formula:
C₈H₄O₃ Boiling Pt: 295 °C (subl.) Melting Pt: 131…133 °C Density: 1.53 Flash Pt: 152 °C (305 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005918 CAS Number: 85-44-9 EINECS: 201-607-5 UN: 2214 ADR: 8,III Merck Index: 14,7372 |
Specification Test Results
Residue after ignition | 0.01% |
Cl | 0.002% |
SO3 | 0.003% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm max. |
Melting point | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...