Phthalic acid ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pathalic acid
41770-22
41770-36
CAAA41770-22EA
64.78
CAD
CAAA41770-22
CAAA41770-36
Phthalic acid ≥99.5% ACS
Phthalic acid
MDL: MFCD00002467
Beilstein Registry No.: 608199
Purity: 99.5+%
CAS No.: 88-99-3
Molecular Formula: C6H4(COOH)2
Formula Weight: 166.13
Flash Point: 168°(334°F)
Physical Form: Crystalline powder
Density: 1.593
Appearance: White
Odor: Odorless
Melting Point: 205° dec.
Beilstein Reference: 608199
Merck Reference: 14,7371
MDL No.: MFCD00002467
Formula:
C₈H₆O₄ MW: 166.13 g/mol Boiling Pt: 315…317 °C (1013 hPa) Melting Pt: 205 °C (dec.) Density: 1.593 Flash Pt: 168 °C (334 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002467 CAS Number: 88-99-3 EINECS: 201-873-2 Merck Index: 14,7371 |
Specification Test Results
Insoluble matter | 0.05% |
Residue after ignition (as SO3) | 0.02% |
Cl | 0.001% |
No | 0.005% |
SO3 | 0.005% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.001% |
H2O | 0.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...