Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

41764-HW 41764-HX
CAAA41764-HWEA 316 CAD
CAAA41764-HW CAAA41764-HX
Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Molybdenum

MDL: MFCD00003465


Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈19.5g/100x150mm, Temper: Hard MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR