Disodium oxalate ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
MDL: MFCD00012465
Beilstein Registry No.: 3631622
Soluble in water. Insoluble in alcohol
Purity: 99.5+%
CAS No.: 62-76-0
Molecular Formula: Na2C2O4
Formula Weight: 134.00
Physical Form: Powder
Density: 2.340
Appearance: White
Odor: Odorless
Melting Point: 250-270° dec.
Solubility: Soluble in water. Insoluble in alcohol
Beilstein Reference: 3631622
Merck Reference: 14,8650
MDL No.: MFCD00012465
Formula:
Na₂C₂O₄ MW: 134 g/mol Melting Pt: 250…270 °C (dec.) Density: 2.340 Storage Temperature: Ambient |
MDL Number:
MFCD00012465 CAS Number: 62-76-0 EINECS: 200-550-3 Merck Index: 14,8650 |
Specification Test Results
Insoluble matter | 0.005% |
Loss on drying at 105°C | 0.01% |
Neutrality | Pass test |
Cl | 0.002% |
SO3 | 0.002% |
NH3 | 0.002% |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.001% |
K | 0.005%% |
Substances darkened by hot sulfuric acid P.T. | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...