Dioctyltin dichloride ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41738-06
41738-14
041738-03
CAAA41738-06EA
96.22
CAD
CAAA41738-06
CAAA41738-14
AA41738-03
Dioctyltin dichloride ≥95%
Dioctyltin dichloride
MDL: MFCD00018091
Purity: 95%
CAS No.: 3542-36-7
Molecular Formula: [CH3(CH2)7]2SnCl2
Formula Weight: 416.04
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Boiling Point: 175°/1mm
UN No.: 3261
MDL No.: MFCD00018091
Formula:
C₁₆H₃₄Cl₂Sn Boiling Pt: 175 °C (1 mmHg) Melting Pt: 44…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018091 CAS Number: 3542-36-7 EINECS: 222-583-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...