Lithium iodide, anhydrous 98+%

Supplier: Thermo Scientific Chemicals
Warning

41733-18 41733-30
CAAA41733-18EA 267.02 CAD
CAAA41733-18 CAAA41733-30
Lithium iodide, anhydrous 98+%
Lithium iodide

As absorbent in refrigeration process, as a catalyst, and in photography


Purity: 98+% CAS No.: 10377-51-2 Molecular Formula: ILi Formula Weight: 133.84 Physical Form: Lumps/powder, Ampouled under argon Density: 4.076 Refractive Index: 1.955 Sensitivity: Light Sensitive and Hygroscopic Appearance: White to pale yellow Odor: Odorless Melting Point: 449° Boiling Point: 1170-1190° Solubility: Soluble in water, alcohol, acetone and methanol. Very soluble in NH4OH Merck Reference: 14,5535 MDL No.: MFCD00011092


As absorbent in refrigeration process, as a catalyst, and in photography

Formula: ILi
Boiling Pt: 1170…1190 °C
Melting Pt: 449 °C
Density: 4.076
Storage Temperature: Ambient
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Merck Index: 14,5535

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR