Cadmium(II) nitrate tetrahydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cadmium nitrate tetrahydrate

41732-18 41732-30
CAAA41732-18EA 211.72 CAD
CAAA41732-18 CAAA41732-30
Cadmium(II) nitrate tetrahydrate ≥99.9% (metals basis)
Cadmium (II) nitrate tetrahydrate

MDL: MFCD00149626


Purity: 99.9% CAS No.: 10022-68-1 Molecular Formula: Cd(NO3)2.4H2O Formula Weight: 308.47 (236.41anhy) Physical Form: Crystalline Density: 2.45 Melting Point: 59° Boiling Point: 132° Merck Reference: 14,1622 UN No.: 3087 MDL No.: MFCD00149626

Formula: CdN₂O₆·4H₂O
MW: 308.48 g/mol
Boiling Pt: 132
Melting Pt: 60
Density: 2.45
Storage Temperature: Ambient
MDL Number: MFCD00149626
CAS Number: 10022-68-1
EINECS: 233-710-6
UN: 3087
ADR: 5.1 II
Merck Index: 14,1622

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR