Chloroacetic acid 99% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Monochloroacetic acid
41724-22
41724-36
41724-57
CAAA41724-22EA
74.42
CAD
CAAA41724-22
CAAA41724-36
CAAA41724-57
Chloroacetic acid 99% ACS
Chloroacetic acid
MDL: MFCD00002683
Beilstein Registry No.: 605438
Purity: 99%
CAS No.: 79-11-8
Molecular Formula: C2H3ClO2
Formula Weight: 94.50
Flash Point: 126°(258°F)
Physical Form: Crystalline
Density: 1.404
Appearance: White
Odor: Pungent
pH:
Formula:
ClCH₂COOH MW: 94.5 g/mol Boiling Pt: 189 °C Melting Pt: 60…63 °C Density: 1.404 Flash Pt: 126 °C (258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002683 CAS Number: 79-11-8 EINECS: 201-178-4 UN: 1751 ADR: 6.1,II Merck Index: 14,2112 |
Specification Test Results
Insoluble matter | 0.01% |
Residue after ignition | 0.02% |
Carbonyl compounds (as acetone) | 0.02% |
Other carbonyl compounds | 0.01% |
Cl | 0.01% |
SO3 | 0.02% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.002% |
Substances darkened by sulfuric acid | Pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...