Sodium diphenylamine-4-sulfonate ACS

Supplier: Thermo Scientific Chemicals
Warning

41714-04 41714-09 41714-18
CAAA41714-04EA 50.24 CAD
CAAA41714-04 CAAA41714-09 CAAA41714-18
Sodium diphenylamine-4-sulfonate ACS
Sodium diphenylamine-4-sulphonate

Redox indicator


CAS No.: 6152-67-6 Molecular Formula: C12H10NNaO3S Formula Weight: 271.27 Physical Form: Powder MDL No.: MFCD00036468


Redox indicator

Formula: C₁₂H₁₀NNaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR