Arsenic(III) telluride 99,999 Metals basis

Supplier: Thermo Scientific Chemicals
Danger

41709-09 41709-18
CAAA41709-09EA 229.1 CAD
CAAA41709-09 CAAA41709-18
Arsenic(III) telluride 99,999 Metals basis
Arsenic (III) telluride

For vacuum deposition


Purity: 99.999% CAS No.: 12044-54-1 Molecular Formula: As2Te3 Formula Weight: 532.64 Physical Form: 3-12mm Pieces Density: 6.50 Melting Point: 381° UN No.: 1557 MDL No.: MFCD00014167


Vacuum deposition

Formula: As₂Te₃
MW: 532.64
Melting Pt: 621
Density: 6.50
Storage Temperature: Ambient
MDL Number: MFCD00014167
CAS Number: 12044-54-1
EINECS: 234-955-1
UN: 1557
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR