Copper(II) oxide ≥99.7% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41692-22
41692-36
41692-A3
CAAA41692-22EA
134.93
CAD
CAAA41692-22
CAAA41692-36
CAAA41692-A3
Copper(II) oxide ≥99.7% (metals basis)
Copper (II) oxide
MDL: MFCD00010979
Purity: 99.7%
CAS No.: 1317-38-0
Molecular Formula: CuO
Formula Weight: 79.55
Physical Form: -200 Mesh Powder
Density: 6.3
Refractive Index: 2.63
Appearance: Black
Odor: Odorless
Melting Point: 1326°
Merck Reference: 14,2646
UN No.: 3077
MDL No.: MFCD00010979
Formula:
CuO Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 1326 °C Density: 6.3 Storage Temperature: Ambient |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9,III Merck Index: 14,2646 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...