Sodium acetate hydrate ≥99.9985% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
41685-06
41685-14
41685-22
CAAA41685-06EA
94.17
CAD
CAAA41685-06
CAAA41685-14
CAAA41685-22
Sodium acetate hydrate ≥99.9985% (metals basis), Puratronic®
Sodium acetate hydrate
MDL: MFCD00071557
Fieser: 1,1024 5,59
Purity: 99.9985%
CAS No.: 41484-91-7
Molecular Formula: NaOOCCH3.xH2O (x~3)
Formula Weight: 82.03(anhy)
Physical Form: Powder
Density: 1.45
Melting Point: 58°
Merck Reference: 14,8571
MDL No.: MFCD00071557
Formula:
H₃CCOONa·3H₂O Melting Pt: 58 °C Density: 1.45 Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 41484-91-7 EINECS: 204-823-8 Merck Index: 14,8571 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...