Terbium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

41678-03 41678-06
CAAA41678-03EA 125.93 CAD
CAAA41678-03 CAAA41678-06
Terbium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Terbium (III) chloride

MDL: MFCD00011258 Soluble in water


Purity: 99.9% CAS No.: 10042-88-3 Molecular Formula: TbCl3 Formula Weight: 265.29 Physical Form: -250 Mesh Powder Sensitivity: Hygroscopic Solubility: Soluble in water Merck Reference: 14,9157 MDL No.: MFCD00011258

Formula: Cl₃Tb
MW: 265.28 g/mol
Melting Pt: 588 °C
Density: 4.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011258
CAS Number: 10042-88-3
EINECS: 233-132-4
Merck Index: 14,9157

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR