Tungsten carbide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41665-18
41665-30
CAAA41665-18EA
284.4
CAD
CAAA41665-18
CAAA41665-30
Tungsten carbide ≥99.5% (metals basis)
Tungsten carbide
MDL: MFCD00801036
Purity: 99.5%
CAS No.: 12070-13-2
Molecular Formula: CW2
Formula Weight: 379.71
Physical Form: -325 Mesh Powder
Density: 17.15
Appearance: Black
Melting Point: ≈2800°
UN No.: 3178
MDL No.: MFCD00801036
Formula:
CW₂ Boiling Pt: 6000 °C (1013 hPa) Melting Pt: ∼2800 °C Density: 17,15 Storage Temperature: Ambient |
MDL Number:
MFCD00801036 CAS Number: 12070-13-2 EINECS: 235-124-6 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...