Calcium ≥98.8% (metals basis), granules -9 mesh

Supplier: Thermo Scientific Chemicals
Danger

41653-18 41653-30 41653-A1
CAAA41653-18EA 79 CAD
CAAA41653-18 CAAA41653-30 CAAA41653-A1
Calcium ≥98.8% (metals basis), granules -9 mesh
Calcium

MDL: MFCD00085314


Purity: 98.8% CAS No.: 7440-70-2 Odor: Odorless UN No.: 1401 MDL No.: MFCD00085314

Formula: Ca
Boiling Pt: 1484 °C (1013 hPa)
Melting Pt: 850 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 213-179-5
UN: 1401
ADR: 4.3,II
Merck Index: 13,01644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR