Strontium titanate S.A. 12.5-25 m²/g S.A. 12.5-25 m²/g

Supplier: Thermo Scientific Chemicals

41634-22 41634-36
CAAA41634-22EA 308.1 CAD
CAAA41634-22 CAAA41634-36
Strontium titanate S.A. 12.5-25 m²/g S.A. 12.5-25 m²/g
Strontium titanate

MDL: MFCD00049554 Notes: Crystalline and spherical particles


CAS No.: 12060-59-2 Molecular Formula: SrTiO3 Formula Weight: 183.51 Physical Form: 0.05-0.15 Micron APS Powder Density: 4.810 Odor: Odorless MDL No.: MFCD00049554

Formula: O₃SrTi
MW: 183.5 g/mol
Melting Pt: 2060 °C
Density: 4,810
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR