Barium titanate 99+% (metals basis), REacton®, high surface area

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

41633-14 41633-22
CAAA41633-14EA 126.4 CAD
CAAA41633-14 CAAA41633-22
Barium titanate 99+% (metals basis), REacton®, high surface area
Barium titanate

MDL: MFCD00003447 Notes: Crystalline and spherical particles


Purity: 99+% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: 0.05-0.07 Micron APS Powder, S.A. 10-30m2/g Density: 5.85 Appearance: White to grey Odor: Odorless Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447

Formula: BaO₃Ti
MW: 233.21 g/mol
Melting Pt: 1620 °C
Density: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,1000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR