Tin ≥98.8% (metals basis), foil, Thickness: 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

41604-CL 41604-VA 41604-VE
CAAA41604-CLEA 1338.26 CAD
CAAA41604-CL CAAA41604-VA CAAA41604-VE
Tin ≥98.8% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Tin

MDL: MFCD00133862


Purity: 98.8% CAS No.: 7440-31-5 Physical Form: ≈8.2g/15x15cm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR