Silver ≥99.9% (metals basis), spherical powder -635 mesh

Supplier: Thermo Scientific Chemicals

41600-30 41600-30 41600-09 41600-09
CAAA41600-30EA 2605.42 CAD
AA41600-30 AA41600-09 CAAA41600-30 CAAA41600-09
Silver ≥99.9% (metals basis), spherical powder -635 mesh
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 961.9 °C
Density: 10.5 g/cm³ (20 °C)
MDL Number: MFCD00003397
CAS Number: 7440-22-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR