Lithium hydride ≥99.4% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
41596-14
41596-22
CAAA41596-14EA
199.08
CAD
CAAA41596-14
CAAA41596-22
Lithium hydride ≥99.4% (metals basis)
Lithium hydride
MDL: MFCD00011074
Fieser: 4,304 13,165 18,217 19,198 20,222
Purity: 99.4%
CAS No.: 7580-67-8
Molecular Formula: HLi
Formula Weight: 7.95
Physical Form: Powder
Density: 0.78
Appearance: White to grey
Odor: Odorless
Melting Point: 686°
Merck Reference: 14,5533
UN No.: 1414
MDL No.: MFCD00011074
Formula:
HLi MW: 7.95 g/mol Melting Pt: 686 °C Density: 0.78 Storage Temperature: Ambient |
MDL Number:
MFCD00011074 CAS Number: 7580-67-8 EINECS: 231-484-3 UN: 1414 ADR: 4.3 I Merck Index: 14,5533 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...