Calcium hydride ≥92%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41590-22
41590-36
CAAA41590-22EA
208.56
CAD
CAAA41590-22
CAAA41590-36
Calcium hydride ≥92%
Calcium hydride
MDL: MFCD00010897
Decomposes in water, alcohol and acids to H2
Fieser: 1,105 18,83
Purity: 92% min
CAS No.: 7789-78-8
Molecular Formula: CaH2
Formula Weight: 42.10
Physical Form: 1-20mm Granules
Density: 1.900
Melting Point: 600° dec.
Solubility: Decomposes in water, alcohol and acids to H2
Merck Reference: 14,1672
UN No.: 1404
MDL No.: MFCD00010897
Formula:
H₂Ca Melting Pt: 600 °C (dec.) Density: 1.900 Storage Temperature: Ambient |
MDL Number:
MFCD00010897 CAS Number: 7789-78-8 EINECS: 232-189-2 UN: 1404 ADR: 4.3,I Merck Index: 14,1672 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...