Tripotassium orthophosphate, anhydrous 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41587-30
41587-A1
41587-A7
CAAA41587-30EA
91.96
CAD
CAAA41587-30
CAAA41587-A1
CAAA41587-A7
Tripotassium orthophosphate, anhydrous 97%
tri-Potassium orthophosphate
MDL: MFCD00036295
Soluble in water. Insoluble in alcohol
Purity: 97%
CAS No.: 7778-53-2
Molecular Formula: K3PO4
Formula Weight: 212.27
Physical Form: Granular
Density: 2.564
Melting Point: 1340°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,7660
MDL No.: MFCD00036295
Formula:
K₃O₄P Melting Pt: 1340 °C Density: 2.564 Storage Temperature: Ambient |
MDL Number:
MFCD00036295 CAS Number: 7778-53-2 EINECS: 231-907-1 Merck Index: 14,7660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...