Tripotassium orthophosphate monohydrate ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
41586-36
41586-A3
CAAA41586-36EA
167.48
CAD
CAAA41586-36
CAAA41586-A3
Tripotassium orthophosphate monohydrate ≥95%
tri-Potassium orthophosphate monohydrate
MDL: MFCD00150410
Purity: 95%
CAS No.: 27176-10-9
Molecular Formula: K3PO4.H2O
Formula Weight: 230.28 (212.27anhy)
Physical Form: Granular
Density: 2.564
MDL No.: MFCD00150410
Formula:
K₃O₄P·1H₂O Density: 2,564 Storage Temperature: Ambient |
MDL Number:
MFCD00150410 CAS Number: 27176-10-9 EINECS: 231-907-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...