Cadmium acetate dihydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
41581-22
41581-36
41581-A3
CAAA41581-22EA
99.7
CAD
CAAA41581-22
CAAA41581-36
CAAA41581-A3
Cadmium acetate dihydrate 98%
Cadmium acetate dihydrate
In cadium electroplating
Purity: 98%
CAS No.: 5743-04-4
Molecular Formula: Cd(OOCCH3)2.2H2O
Formula Weight: 266.54 (230.50anhy)
Physical Form: Powder
Density: 2.01
Appearance: White
Solubility: Freely soluble in water, soluble in alcohol
Merck Reference: 14,1614
UN No.: 2570
MDL No.: MFCD00012449
In cadium electroplating
Formula:
(H₃CCOO)₂Cd·2H₂O Boiling Pt: 105…107 °C (4 mmHg) Melting Pt: 256 °C Density: 2.01 Storage Temperature: Ambient |
MDL Number:
MFCD00012449 CAS Number: 5743-04-4 EINECS: 208-853-2 UN: 2570 ADR: 6.1,III Merck Index: 14,1614 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...