Antimony(III) bromide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
41573-18
41573-30
CAAA41573-18EA
251.22
CAD
CAAA41573-18
CAAA41573-30
Antimony(III) bromide 99%
Antimony (III) bromide
MDL: MFCD00016317
Purity: 99%
CAS No.: 7789-61-9
Molecular Formula: SbBr3
Formula Weight: 361.48
Physical Form: Powder
Density: 4.148
Refractive Index: 1.74
Sensitivity: Moisture Sensitive
Odor: Characteristic
Melting Point: 96.6°
Boiling Point: 280°
Merck Reference: 14,706
UN No.: 3260
MDL No.: MFCD00016317
Formula:
Br₃Sb Boiling Pt: 280 °C Melting Pt: 96.6 °C Density: 4.148 Storage Temperature: Ambient |
MDL Number:
MFCD00016317 CAS Number: 7789-61-9 EINECS: 232-179-8 UN: 3260 ADR: 8,II Merck Index: 14,706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...