Calcium hexaboride 99+% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41551-14
41551-22
CAAA41551-14EA
159.58
CAD
CAAA41551-14
CAAA41551-22
CAAA41551-36
Calcium hexaboride 99+% (metals basis)
Calcium hexaboride
MDL: MFCD00148904
Purity: 99+%
CAS No.: 12007-99-7
Molecular Formula: CaB6
Formula Weight: 104.95
Physical Form: -20 Mesh Powder
Density: 2.30
Melting Point: 2235°
MDL No.: MFCD00148904
Formula:
B₆Ca Melting Pt: 2235 °C Density: 2,30 Storage Temperature: Ambient |
MDL Number:
MFCD00148904 CAS Number: 12007-99-7 EINECS: 234-525-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...