Potassium sulfide (K2Sx) (≥11.8% S) (as sulfide)
Supplier: Thermo Scientific Chemicals
|
Danger
|
41550-22
41550-36
CAAA41550-22EA
65.73
CAD
CAAA41550-22
CAAA41550-36
Potassium sulfide (K2Sx) (≥11.8% S) (as sulfide)
Potassium sulphide (K2Sx)
Decorative color effects on brass, bronze, nickel
CAS No.: 39365-88-3
Molecular Formula: K2Sn
Sensitivity: Air Sensitive and Hygroscopic
Solubility: Soluble in water
UN No.: 3180
MDL No.: MFCD00064604
Decorative color effects on brass, bronze, nickel
Formula:
K₂(S)n Melting Pt: 471 °C Density: 1.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00148074 CAS Number: 37199-66-9 EINECS: 253-390-1 UN: 3180 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...