(±)-1-Methoxy-2-propanol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Monopropylene glycol methyl ether

41547-AK 41547-K2 41547-K7 41547-M6
CAAA41547-AKEA 50.4 CAD
CAAA41547-AK CAAA41547-K2 CAAA41547-K7 CAAA41547-M6
(±)-1-Methoxy-2-propanol 99+%
(±)-1-Methoxy-2-propanol

MDL: MFCD00004537 Beilstein Registry No.: 1731270


Purity: 99+% CAS No.: 107-98-2 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 0.926 Refractive Index: 1.4030 Appearance: Colorless Odor: Alcohol-like pH: 4-7 Melting Point: -100° Boiling Point: 119-120° Beilstein Reference: 1731270 UN No.: 3092 MDL No.: MFCD00004537

Formula: CH₃CH(OH)CH₂OCH₃
Boiling Pt: 119…120 °C
Melting Pt: –100 °C
Density: 0.926
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004537
CAS Number: 107-98-2
EINECS: 203-539-1
UN: 3092
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e076d8e7a4ef95472e37d2c355f37d64eac8ee&vid=26da88ae9f09b8195abc9bdd949f6bbdaf429255&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR