Titanium ≥99.9% (metals basis), spherical powder -150 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
41545-14
41545-22
41545-36
CAAA41545-14EA
79.47
CAD
CAAA41545-14
CAAA41545-22
CAAA41545-36
Titanium ≥99.9% (metals basis), spherical powder -150 mesh
Titanium
MDL: MFCD00011264
Purity: 99.9%
CAS No.: 7440-32-6
Odor: Odorless
UN No.: 2546
MDL No.: MFCD00011264
Formula:
Ti Boiling Pt: 3277 °C (1013 hPa) Melting Pt: 1677 °C Density: 4,5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 2546 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...