Sulfanilic acid 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
41540-18
41540-30
CAAA41540-18EA
66.2
CAD
CAAA41540-18
CAAA41540-30
Sulfanilic acid 98.0-102.0% ACS
Sulphanilic acid
Reagent for chromatographic detection of histidine
Purity: 98.0-102.0%
CAS No.: 121-57-3
Molecular Formula: C6H7NO3S
Formula Weight: 173.19
Physical Form: Powder
Melting Point: ca 365°
Beilstein Reference: 908765
Merck Reference: 14,8926
MDL No.: MFCD00007886
Reagent for chromatographic detection of histidine
Formula:
4-(H₂N)C₆H₄SO₃H Boiling Pt: 363 °C (1013 hPa) Melting Pt: ∼365 °C Density: 1.485 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007886 CAS Number: 121-57-3 EINECS: 204-482-5 Merck Index: 14,8926 |
|
Specification Test Results
Residue after ignition | 0.01% |
Insoluble in sodium carbonate | 0.02% |
Cl | 0.002% |
NO3 | 0.5ppm |
SO3 | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...