Antimony(III) chloride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
41537-14
41537-22
CAAA41537-14EA
103.33
CAD
CAAA41537-14
CAAA41537-22
Antimony(III) chloride ≥99.9% (metals basis)
Antimony (III) chloride
MDL: MFCD00011212
Fieser: 8,19 18,25 19,14
Purity: 99.9%
CAS No.: 10025-91-9
Molecular Formula: Cl3Sb
Formula Weight: 228.11
Physical Form: Crystalline
Density: 3.14
Appearance: White
Odor: Pungent
Melting Point: 73.4°
Boiling Point: 223.5°
Merck Reference: 14,707
UN No.: 1733
MDL No.: MFCD00011212
Formula:
SbCl₃ Boiling Pt: 223.5 °C Melting Pt: 73.4 °C Density: 3.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011212 CAS Number: 10025-91-9 EINECS: 233-047-2 UN: 1733 ADR: 8,II Merck Index: 14,707 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...