Sodium metaarsenite standardised solution 0.1 N in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

41533-AP 41533-K7
CAAA41533-APEA 232.26 CAD
CAAA41533-AP CAAA41533-K7
Sodium metaarsenite standardised solution 0.1 N in aqueous solution
Sodium metaarsenite

MDL: MFCD00003472


CAS No.: 7784-46-5 Molecular Formula: AsNaO2 Formula Weight: 129.91 Physical Form: Liquid Appearance: Colorless Odor: Odorless UN No.: 1686 MDL No.: MFCD00003472

Formula: NaAsO₂
Storage Temperature: Ambient
MDL Number: MFCD00003472
CAS Number: 7784-46-5
EINECS: 232-070-5
UN: 1686
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR