Holmium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic

Supplier: Thermo Scientific Chemicals
Danger

41515-04 41515-09
CAAA41515-04EA 116.29 CAD
CAAA41515-04 CAAA41515-09
Holmium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Holmium

MDL: MFCD00011049


Purity: 99.9% CAS No.: 7440-60-0 Physical Form: Packaged under argon Appearance: Dark gray to silver Odor: Odorless UN No.: 3178 MDL No.: MFCD00011049

Formula: Ho
MW: 164.93
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0
UN: 3178
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR