Trifluoroiodomethane 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
41479-14
41479-22
CAAA41479-14EA
606.72
CAD
CAAA41479-14
CAAA41479-22
Trifluoroiodomethane 97+%
Trifluoroiodomethane
MDL: MFCD00001060
Beilstein Registry No.: 1732740
Purity: 97+%
CAS No.: 2314-97-8
Molecular Formula: CF3I
Formula Weight: 195.91
Flash Point: None
Physical Form: Gas, Packaged in cylinders
Density: 2.361
Refractive Index: 1.3790
Sensitivity: Light Sensitive
Boiling Point: -22.5°
Beilstein Reference: 1732740
UN No.: 1956
MDL No.: MFCD00001060
Formula:
CF₃I Boiling Pt: –22,5 °C Melting Pt: –110 °C Density: 2,361 Storage Temperature: Ambient |
MDL Number:
MFCD00001060 CAS Number: 2314-97-8 EINECS: 219-014-5 UN: 1956 ADR: 2.2, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...