tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41470-AK
41470-K2
41470-K7
CAAA41470-AKEA
82.95
CAD
CAAA41470-AK
CAAA41470-K2
CAAA41470-K7
tert-Butanol, anhydrous ≥99.5%
tert-Butanol
Solvent in Sol-Gel precursor applications
Purity: 99.5%
CAS No.: 75-65-0
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3870
Appearance: Colorless
Melting Point: 25°
Boiling Point: 83°
Beilstein Reference: 906698
Merck Reference: 14,1542
UN No.: 1120
MDL No.: MFCD00004464
Solvent in Sol-Gel precursor applications
Formula:
(CH₃)₃COH Boiling Pt: 83 °C Melting Pt: 25 °C Density: 0.786 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004464 CAS Number: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3,II Merck Index: 14,1542 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...