Cyclopentene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41461-AC
41461-AE
CAAA41461-ACEA
65.1
CAD
CAAA41461-AC
CAAA41461-AE
Cyclopentene 98+%
Cyclopentene
MDL: MFCD00001394
Beilstein Registry No.: 635707
Purity: 98+%
CAS No.: 142-29-0
Molecular Formula: C5H8
Formula Weight: 68.12
Flash Point: -29°(-20°F)
Physical Form: Liquid
Density: 0.773
Refractive Index: 1.4225
Sensitivity: Air Sensitive
Appearance: Colorless
Melting Point: -135°
Boiling Point: 44-45°
Beilstein Reference: 635707
UN No.: 2246
MDL No.: MFCD00001394
Formula:
C₅H₈ MW: 68.12 g/mol Boiling Pt: 44…45 °C Melting Pt: –135 °C Density: 0,773 Flash Pt: –29 °C (–20 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001394 CAS Number: 142-29-0 EINECS: 205-532-9 UN: 2246 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...