Tripropyltin chloride ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41459-03
41459-06
41459-14
CAAA41459-03EA
88.95
CAD
CAAA41459-03
CAAA41459-06
CAAA41459-14
Tripropyltin chloride ≥95%
Tripropyltin chloride
MDL: MFCD00013610
Purity: 95%
CAS No.: 2279-76-7
Molecular Formula: (CH3CH2CH2)3SnCl
Formula Weight: 283.41
Physical Form: Liquid
Density: 1.2678
Refractive Index: 1.4910
Sensitivity: Moisture Sensitive
Melting Point: -23.5°
Boiling Point: 123°/13mm
UN No.: 2788
MDL No.: MFCD00013610
Formula:
C₉H₂₁ClSn Boiling Pt: 123 °C (13 mmHg) Melting Pt: –23.5 °C Density: 1,2678 Flash Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013610 CAS Number: 2279-76-7 EINECS: 218-910-3 UN: 2788 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...