Silver 99,9 Metals basis, wire, hard, Ø 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
41457-BW
41457-CD
CAAA41457-BWEA
224.36
CAD
CAAA41457-BW
CAAA41457-CB
CAAA41457-CD
Silver 99,9 Metals basis, wire, hard, Ø 2.0 mm (0.08 in)
Silver
MDL: MFCD00003397
Purity: 99.9%
CAS No.: 7440-22-4
Physical Form: ≈32.94g/m, Temper: hard
Odor: Characteristic
MDL No.: MFCD00003397
Formula:
Ag MW: 107.87 g/mol Boiling Pt: 2212 Melting Pt: 962 Density: 10.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...