Trichloro(methyl)germane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloro(methyl)germane, Methylgermanium trichloride

41449-02 41449-04 41449-09
CAAA41449-02EA 69.99 CAD
CAAA41449-02 CAAA41449-04 CAAA41449-09
Trichloro(methyl)germane 97%
Trichloro(methyl)germane

MDL: MFCD00013585


Purity: 97% CAS No.: 993-10-2 Molecular Formula: CH3Cl3Ge Formula Weight: 193.99 Flash Point: 10°(50°F) Physical Form: Liquid Density: 1.706 Refractive Index: 1.4685 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Odor: Chlorine-like Boiling Point: 111° UN No.: 2924 MDL No.: MFCD00013585

Formula: CH₃Cl₃Ge
MW: 194
Boiling Pt: 111 °C
Density: 1.706
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013585
CAS Number: 993-10-2
EINECS: 213-604-6
UN: 2924
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR