18-Pentatriacontanone tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
41445-14
41445-22
CAAA41445-14EA
100.8
CAD
CAAA41445-14
CAAA41445-22
18-Pentatriacontanone tech. 85%, Technical Grade
18-Pentatriacontanone
Powder
Purity: 97+%
CAS No.: 504-53-0
Molecular Formula: C35H70O
Formula Weight: 506.94
Physical Form: Powder
Appearance: Off-white
Melting Point: 79-80°
Solubility: Soluble in benzene
MDL No.: MFCD00048491
Antiblocking agent
Formula:
C₃₅H₇₀O Boiling Pt: 543 °C (1013 hPa) Melting Pt: 79…80 °C Density: 0.84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00048491 CAS Number: 504-53-0 EINECS: 207-993-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...