Silver sulfate ≥99.99% (metals basis) Ag ≥ 68.9%, Premion®

Supplier: Thermo Scientific Chemicals
Danger

41443-14 41443-22
CAAA41443-14EA 227.52 CAD
CAAA41443-14 CAAA41443-22
Silver sulfate ≥99.99% (metals basis) Ag ≥ 68.9%, Premion®
Silver sulphate

MDL: MFCD00003407


Purity: 99.99% CAS No.: 10294-26-5 Molecular Formula: Ag2O4S Formula Weight: 311.80 Physical Form: Powder Density: 5.45 Appearance: White Odor: Odorless Melting Point: 657° Boiling Point: 1085° dec. Merck Reference: 14,8529 UN No.: 3077 MDL No.: MFCD00003407

Formula: Ag₂SO₄
Boiling Pt: 1085 °C (dec.)
Melting Pt: 657 °C
Density: 5.45
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 3077
ADR: 9,III
Merck Index: 14,8529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR