Ytterbium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ytterbium trifluoride, Ytterbium fluoride
41423-09
41423-18
CAAA41423-09EA
221.2
CAD
CAAA41423-09
CAAA41423-18
Ytterbium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Ytterbium (III) fluoride
For vacuum deposition
Purity: 99.9%
CAS No.: 13760-80-0
Molecular Formula: YbF3
Formula Weight: 230.04
Physical Form: 3-12mm Pieces
Density: 8.2
Melting Point: 1157°
UN No.: 3288
MDL No.: MFCD00049615
For vacuum deposition
Formula:
F₃Yb MW: 230.04 Boiling Pt: 2227 Melting Pt: 1157 °C Density: 8.2 Storage Temperature: Ambient |
MDL Number:
MFCD00049615 CAS Number: 13760-80-0 EINECS: 237-354-2 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...