Lanthanum(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lanthanum trifluoride

41414-09 41414-18
CAAA41414-09EA 112.81 CAD
CAAA41414-09 CAAA41414-18
Lanthanum(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Lanthanum (III) fluoride

For vacuum deposition


Purity: 99.9% CAS No.: 13709-38-1 Molecular Formula: F3La Formula Weight: 195.90 Physical Form: 3-6mm Pieces Density: 5.936 Melting Point: 1493° MDL No.: MFCD00011073


For vacuum deposition

Formula: F₃La
MW: 195.9 g/mol
Boiling Pt: 2327
Melting Pt: 1493 °C
Density: 5.936
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS Number: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR