Holmium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Holmium (III) fluoride

41413-04 41413-09
CAAAA41413-04EA 80.74 CAD
CAAAA41413-04 CAAA41413-09 CAAA41413-18
Holmium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Holmium trifluoride

Anhydrous


For vacuum deposition


For vacuum deposition

Formula: HoF₃
MW: 221.93 g/mol
Melting Pt: 1143
Density: 7.6
Storage Temperature: Ambient
MDL Number: MFCD00016142
CAS Number: 13760-78-6
EINECS: 237-352-1
UN: 3288
ADR: 6.1 III

Order Now

Specification Test Results

Form 3-6mm Pieces


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR