Gadolinium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

41412-09 41412-18
CAAA41412-09EA 176.96 CAD
CAAA41412-09 CAAA41412-18
Gadolinium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Gadolinium trifluoride

For vacuum deposition


Purity: 99.9% CAS No.: 13765-26-9 Molecular Formula: GdF3 Formula Weight: 214.25 Physical Form: 3-6mm Pieces Density: 7.1 Appearance: White Odor: Odorless Melting Point: 1231° MDL No.: MFCD00016113


For vacuum deposition

Formula: F₃Gd
Melting Pt: 1231 °C
Density: 7.1
Storage Temperature: Ambient
MDL Number: MFCD00016113
CAS Number: 13765-26-9
EINECS: 237-369-4
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR